missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ SUBHT High Throughput Submarine Gel Unit DFS Item

Ideal for high throughput screening applications like single strand polymorphism (SSP) analysis following PCR

Supplier:  Hoefer™ SUBHTCS

 View more versions of this product

Catalog No. 03-500-407


Only null left
Explore available promotions

Includes:
  • Buffer chamber assembly
  • Safety lid, high-voltage leads
  • Fluorescent Labeled Running Tray with Casting Gates
  • Four 1mm thick 30-well combs
  • Colored Loading Strips
  • Two Buffer Recirculation Ports

Description

Description

  • Fast, error-free loading: load an entire 96-well plate using a multichannel pipetter
  • Simplified Analysis: Fluorescent labels for each sample printed on a UV transparent gel tray allows for instant sample identification (matched to a 96-well plate) during post-run visualization
  • High Throughput: Run up to 120 samples (96-well plate plus 24 control and/or marker samples) in 30 minutes
  • Better Reproducibility: Eliminate variability caused by running buffer pH drift and ionic gradient formation with optional buffer recirculation capability
  • Reduced Buffer Needs: Compact tank design needs less buffer to cover gel which not only reduces buffer consumption but provides greater control of voltage gradients and run times
  • Maximum Flexibility: Fully compatible with a wide range of combs and accessories to meet all your high throughput and standard gel analysis needs
  • This high throughput horizontal gel unit can accommodate up to 120 samples and resolve DNA strands from 50-1000 base pairs in 30 minutes
Specifications
  • Gel Dimensions: 15 x 15 cm
  • Maximum Buffer Volume: 1.2L
  • Maximum Power Settings: 300V, 200mA, 60W
  • Maximum Sample Capacity: 120
  • Maximum Running Temperature: 45°C
  • Indoor Use: 4-40°C
  • Humidity: up to 80%
  • Unit Dimensions: 33.5L x 21.5W x 7 cmH
  • Safety Certifications: EN61010-1, CE

Applications:

Single Strand Polymorphism (SSP) analysis

Specifications

Specifications

SUBHT High Throughput Submarine Gel Unit
Colored Loading Strip
Buffer chamber assembly, safety lid with high voltage Leads, fluorescent labeled running tray with casting gates, four 1mm thick 30-well combs, colored loading strips, two Buffer recirculation ports
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions