missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Suberoyl-L-Carnitine Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™
$2460.88 - $2460.88
Specifications
| CAS | 102636-81-7 (Free Acid) |
|---|---|
| Molecular Formula | C15H27NO6 · xLi+ |
| Linear Formula | C15H27NO6 · xLi+ |
| Optical Rotation | [α]0.1/D -18 ±2° in H2O |
| Formula Weight | 317.38 (Free Acid Basis) |
Specifications
| 102636-81-7 (Free Acid) | |
| C15H27NO6 · xLi+ | |
| 317.38 (Free Acid Basis) | |
| Analytical Standard |
| C15H27NO6 · xLi+ | |
| [α]0.1/D -18 ±2° in H2O | |
| ≥93% (HPLC) |