missing translation for 'onlineSavingsMsg'
Learn More

Suberoyl-L-Carnitine Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022424
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022424 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022424 Supplier Merck Emd Millipore Supplier No. 1586850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 102636-81-7 (Free Acid)
Molecular Formula C15H27NO6 · xLi+
Linear Formula C15H27NO6 · xLi+
Quantity 50 mg
Optical Rotation [α]0.1/D -18 ±2° in H2O
Formula Weight 317.38 (Free Acid Basis)
Percent Purity ≥93% (HPLC)
Grade Analytical Standard