Learn More
Hoefer™ SUB10 Mini-Plus Submarine Combs
Accessories for SUB10 Mini-Plus Submarine
Supplier: Hoefer™ SUB10C1.512
Description
- Ranges from 8, 10, 12, 16 and 20
- 1, 1.5 and 2 mm Thickness
- Size ranges from 3 to 9 mm Well Width
- Min. 12, max. 80 μL Sample Volume
Specifications
| SUB10 Mini plus Submarine Unit | |
| 12 | |
| 0.15 cm | |
| 0.21 in. | |
| 0.19 in. |
| Comb | |
| 0.05 in. | |
| 35 μL | |
| 0.55 cm | |
| 0.5 cm |