missing translation for 'onlineSavingsMsg'
Learn More

Styrene (stabilized with TBC) 99.0+%, TCI America™

Catalog No. S009525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S009525ML 25 mL
S0095500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S009525ML Supplier TCI America Supplier No. S009525ML

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name ethenylbenzene
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31°C
Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 2055
Formula Weight 104.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Styrene (stabilized with TBC)
EINECSNumber (3)-0004
RTECSNumber WL3675000
TSCA Yes
Recommended Storage Refrigerator