missing translation for 'onlineSavingsMsg'
Learn More

Styrene, MilliporeSigma™ Supelco™

Catalog No. 111017131
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017131 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017131 Supplier Merck Emd Millipore Supplier No. 47745U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Styrene is a volatile compound, used in the manufacture of numerous types of plastic, glass fiber-reinforced resins, protective coatings, synthetic rubber and ion exchange resins.

Environmental

Specifications

Chemical Name or Material Styrene
Packaging Ampule
CAS 100-42-5
Ignition Point 914°F
Vapor Pressure 4.3 mmHg (15°C)
Vapor Density 3.6 (vs. air)
Density 0.906 g/mL (at 25°C)
Flash Point 32°C
Physical Form Neat
Linear Formula C6H5 CH=CH2
Technique HPLC
MDL Number MFCD00008612
Melting Point -31°C
Boiling Point 145°C to 146°C
Synonym Phenylethylene; Vinylbenzene
Type Organic Pollutant Standard
Industry Type Environmental Testing
Molecular Weight (g/mol) 104.15
Quantity 1000 mg
Formula Weight 104.15
Grade Analytical Standard
Show More Show Less