missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Styrene, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Petroleum
Specifications
Specifications
| Chemical Name or Material | Styrene |
| CAS | 100-42-5 |
| Ignition Point | 914°F |
| Vapor Pressure | 4.3 mmHg (15°C) |
| Vapor Density | 3.6 (vs. air) |
| Density | 0.906 g/mL (at 25°C) |
| Flash Point | 32°C |
| Physical Form | Neat |
| Linear Formula | C6H5 CH=CH2 |
| Technique | Gas Chromatography |
| Show More |