missing translation for 'onlineSavingsMsg'
Learn More

Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol

CAS: 100-42-5 | C8H8 | 104.15 g/mol

Supplier:  Thermo Scientific Chemicals A18481AP

 View more versions of this product

Catalog No. AAA18481AP


Only null left
Explore available promotions

Description

Description

Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

Solubility
Miscible with carbon disulfide, ether, methanol, benzene, toluene, ethanol, n-heptane, carbon tetrachloride and acetone. Immiscible with water.

Notes
Air and light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and copper.
Specifications

Specifications

Styrene
100-42-5
0.906
145°C to 146°C
Pungent
1.546
500 mL
1071236
14,8860
Miscible with carbon disulfide,ether,methanol,benzene,toluene,ethanol,n-heptane,carbon tetrachloride and acetone. Immiscible with water.
C=CC1=CC=CC=C1
104.15
CHEBI:27452
99%
stabilized with 10 to 15ppm 4-tert-butylcatechol
-31°C
Neutral
31°C (88°F)
C8H8
MFCD00008612,MFCD00084450
UN2055
Light sensitive
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PPBRXRYQALVLMV-UHFFFAOYSA-N
styrene
7501
104.15

Chemical Identifiers

100-42-5
104.15
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
C=CC1=CC=CC=C1
C8H8
MFCD00008612,MFCD00084450
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED

EINECSNumber : 202-851-5

RTECSNumber : WL3675000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only