missing translation for 'onlineSavingsMsg'
Learn More

Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol

Catalog No. AAA18481AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA18481AE 100 mL
AAA18481AP 500 mL
AAA184810F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA18481AE Supplier Thermo Scientific Chemicals Supplier No. A18481AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-42-5 | C8H8 | 104.15 g/mol

Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

Solubility
Miscible with carbon disulfide, ether, methanol, benzene, toluene, ethanol, n-heptane, carbon tetrachloride and acetone. Immiscible with water.

Notes
Air and light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and copper.

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
SMILES C=CC1=CC=CC=C1

Specifications

Melting Point -31°C
Density 0.906
pH Neutral
Boiling Point 145°C to 146°C
Flash Point 31°C (88°F)
Odor Pungent
Refractive Index 1.546
Quantity 100 mL
UN Number UN2055
Beilstein 1071236
Sensitivity Light sensitive
Merck Index 14,8860
Solubility Information Miscible with carbon disulfide,ether,methanol,benzene,toluene,ethanol,n-heptane,carbon tetrachloride and acetone. Immiscible with water.
IUPAC Name styrene
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material Styrene, stabilized with 10 to 15ppm 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372
Hazard Statement GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED
EINECSNumber 202-851-5
RTECSNumber WL3675000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only