Learn More
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol
Description
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.
Solubility
Miscible with carbon disulfide, ether, methanol, benzene, toluene, ethanol, n-heptane, carbon tetrachloride and acetone. Immiscible with water.
Notes
Air and light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and copper.
Specifications
Specifications
| Melting Point | -31°C |
| Density | 0.906 |
| pH | Neutral |
| Boiling Point | 145°C to 146°C |
| Flash Point | 31°C (88°F) |
| Odor | Pungent |
| Refractive Index | 1.546 |
| Quantity | 2500 mL |
| UN Number | UN2055 |
| Beilstein | 1071236 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372 |
| Hazard Statement | GHS H Statement H372-H304-H361-H226-H332-H315-H319-H335 Causes damage to organs through prolonged or repeated exposure. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED |
| EINECSNumber | 202-851-5 |
| RTECSNumber | WL3675000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only