missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Styrene

Catalog No. S4972165L
Encompass
Change view
Click to view available options
Quantity:
16.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S4972165L 16.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S4972165L Supplier Sigma Aldrich Supplier No. S497216.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Styrene

Specifications

CAS 100-42-5
Melting Point -31°C
Density 0.906 g/mL at 25°C
Boiling Point 145°C to 146°C
Molecular Formula C8H8
Linear Formula C6H5CH=CH2
Refractive Index n20/D 1.546
MDL Number MFCD00008612
Quantity 16.5 L
Synonym Phenylethylene; Vinylbenzene
Molecular Weight (g/mol) 104.15 g/mol
Show More Show Less