missing translation for 'onlineSavingsMsg'
Learn More

Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%

Catalog No. AAL1168506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1168506 5 g
AAL1168514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1168506 Supplier Thermo Scientific Chemicals Supplier No. L1168506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2695-37-6 | C8H7NaO3S | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2695-37-6
Molecular Formula C8H7NaO3S
Molecular Weight (g/mol) 206.19
MDL Number MFCD00013379 MFCD00084449
InChI Key XFTALRAZSCGSKN-UHFFFAOYSA-M
Synonym sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt
PubChem CID 3571582
IUPAC Name sodium;4-ethenylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1

Specifications

Melting Point >320°C
Quantity 5 g
Beilstein 3575931
Formula Weight 206.20 (Anhydrous)
Percent Purity 90%
Grade Technical
Chemical Name or Material Styrene-4-sulfonic acid sodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-266-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only