missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX7 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA5149012
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.Chemical Identifiers
| STX7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human, Mouse, Rat | |
| Stx7 | |
| AI315064; AI317144; AW107239; STX7; Syn7; syntaxin 7; Syntaxin7; syntaxin-7; syntaxin-7-like; syntaxin-7-like protein; Unknown (protein for MGC:128117) | |
| Stx7 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Sheep | |
| RUO | |
| PBS with trehalose and no preservative | |
| O15400, O70257, O70439 | |
| 53331, 60466, 8417 | |
| E. coli-derived recombinant human Syntaxin 7, Asn21-Glu187. | |
| Antibody |
Specifications
| STX7 | |
| Polyclonal | |
| Unconjugated | |
| Stx7 | |
| AI315064; AI317144; AW107239; STX7; Syn7; syntaxin 7; Syntaxin7; syntaxin-7; syntaxin-7-like; syntaxin-7-like protein; Unknown (protein for MGC:128117) | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 53331, 60466, 8417 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O15400, O70257, O70439 | |
| Stx7 | |
| E. coli-derived recombinant human Syntaxin 7, Asn21-Glu187. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |