missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565939
Description
Immunogen sequence: KERLMNDFSAA LNNFQAVQRR VSEKEKESIA RARAGSRLSA EERQREEQLV SFDSHEEWNQ MQSQEDEVAI TEQDLELIKE RET Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).Chemical Identifiers
| STX12 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| STX12 | |
| AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13 | |
| STX12 | |
| Primary | |
| Antigen affinity chromatography |
| 0.20 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y82 | |
| 23673 | |
| Recombinant Human STX12. Recombinant protein control fragment (Product #RP-105013). | |
| Antibody |
Specifications
| STX12 | |
| Polyclonal | |
| Unconjugated | |
| STX12 | |
| AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86Y82 | |
| STX12 | |
| Recombinant Human STX12. Recombinant protein control fragment (Product #RP-105013). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |