Learn More
Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL
Sr(NO3)2 in 5% HNO3
$99.58 - $308.93
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA13874AD
|
Thermo Scientific Chemicals
013874AD |
50 mL |
Each for $99.58
|
|
|||||
|
AA13874AE
|
Thermo Scientific Chemicals
013874AE |
100 mL |
Each for $134.88
|
|
|||||
|
AA13874AP
|
Thermo Scientific Chemicals
013874AP |
500 mL |
Each for $308.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Sr(NO3)2 in 5% HNO3 | |
| MFCD00134060 |
Specifications
| 23 hPa (17mm Hg) at 20°C | |
| Sr(NO3)2 in 5% HNO3 | |
| 50 mL | |
| Strontium, plasma standard solution, Sr 1000μg/mL |
| Liquid | |
| MFCD00134060 | |
| Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76% |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8; Packaging Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only