missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxide, tech.

Catalog No. p-7049017
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1249530 250 g
AA12495A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1249530 Supplier Thermo Scientific Chemicals Supplier No. 01249530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1314-11-0 | OSr | 103.619 g/mol

In the manufacture of strontium salts, pyrotechnics, pigments, in greases and soaps and as a desiccant.Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industries. It is an active component of paint dryers. It is also used to prepare pure strontium upon heating with aluminum under vacuum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant

Chemical Identifiers

CAS 1314-11-0
Molecular Formula OSr
Molecular Weight (g/mol) 103.619
MDL Number MFCD00049558
InChI Key UFQXGXDIJMBKTC-UHFFFAOYSA-N
PubChem CID 518709
IUPAC Name oxostrontium
SMILES O=[Sr]

Specifications

Boiling Point ∼3000°C
Melting Point 2430°C
Physical Form Powder
Quantity 250 g
UN Number UN3262
Merck Index 14,8846
Solubility Information Soluble in fused potassium hydroxide; Insoluble in ether and acetone; Decomposes in water to Sr(OH)2 with evolution of heat;Slightly soluble in alcohol; Insoluble in acetone and ether.
Formula Weight 103.62
Grade Technical
Odor Odorless
Sensitivity Moisture sensitive
Density 4.700 g/mL
Chemical Name or Material Strontium oxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.
EINECSNumber 215-219-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only