missing translation for 'onlineSavingsMsg'
Learn More

Strontium oxalate, 95%, Thermo Scientific Chemicals

Catalog No. AA3957336
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3957336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3957336 Supplier Thermo Scientific Chemicals Supplier No. 03957336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 814-95-9 | Molecular Formula: C2O4Sr | Molecular Weight: 175.638 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 814-95-9
Molecular Formula C2O4Sr
Molecular Weight (g/mol) 175.638
MDL Number MFCD00050803
InChI Key KQAGKTURZUKUCH-UHFFFAOYSA-L
Synonym strontium oxalate, unii-7iaw7e69ni, 7iaw7e69ni, strontium oxalate, src2o4, ethanedioic acid, strontium salt 1:1, oxalic acid, monostrontium salt, strontium 2+ ion oxalate, oxalic acid, strontium salt 1:1 8ci
PubChem CID 69947
IUPAC Name strontium;oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Sr+2]

Specifications

Quantity 500 g
UN Number UN3288
Solubility Information Soluble in hydrochloric acid and nitric acid
Formula Weight 175.64
Percent Purity 95%
Physical Form Powder or Lumps
Chemical Name or Material Strontium oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 212-415-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only