missing translation for 'onlineSavingsMsg'
Learn More

Strontium, Oil based standard solution, Specpure™ Sr 1000μg/g

Catalog No. AA1420518
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1420518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1420518 Supplier Thermo Scientific Chemicals Supplier No. 01420518
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00134060

Specifications

Physical Form Liquid
Quantity 50 g
Grade Specpure
Chemical Name or Material Strontium Oil based standard solution, Sr 1000μg/g
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only