missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-4407967
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4319314 25 g
AA4319322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4319314 Supplier Thermo Scientific Chemicals Supplier No. 04319314
Only null left

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

Used in the manufacture of lights, flares, flame tube, the TV picture tube with the red light emitting agent, red fireworks, optical glass and phosphor. It is used as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Soluble in water,liquid ammonia. Slightly soluble in ethanol and acetone. Insoluble in nitric acid.
Formula Weight 211.63
Sensitivity Hygroscopic
Density 2.99
Chemical Name or Material Strontium nitrate
Hazard Category H272-H318
Precautionary Statement P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 233-131-9
RTECSNumber WK9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only