missing translation for 'onlineSavingsMsg'
Learn More

Strontium nitrate, Puratronic™, 99.9965% (metals basis)

Catalog No. p-4407966
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1087914 25 g
AA1087922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1087914 Supplier Thermo Scientific Chemicals Supplier No. 1087914
Only null left

CAS: 10042-76-9 | N2O6Sr | 211.63 g/mol

Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. It mixed with glycolic acid and used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 570°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9965% (metals basis)
Solubility Information Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid.
IUPAC Name strontium(2+) dinitrate
Formula Weight 211.63
Sensitivity Hygroscopic
Density 2.99 g/mL
Chemical Name or Material Strontium nitrate
Hazard Category H272-H318
Precautionary Statement P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 233-131-9
RTECSNumber WK9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only