missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S168025KGPL
Specifications
| 10042-76-9 | |
| 5.0 to 7.0 | |
| 99% | |
| 0.001 |
| 1 | |
| 2.5 kg | |
| ACS | |
| Poly Pail |
Chemical Identifiers
| 10042-76-9 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.