missing translation for 'onlineSavingsMsg'
Learn More

Strontium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1680500GM

 View more versions of this product

Catalog No. 18614113

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10042-76-9
5.0 to 7.0
99%
0.001
1
500 g
ACS
Poly Bottle

Chemical Identifiers

10042-76-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.