missing translation for 'onlineSavingsMsg'
Learn More

Strontium molybdenum oxide, 99%

CAS: 13470-04-7 | SrMoO4

Supplier:  Thermo Scientific Chemicals 040238A1

 View more versions of this product

Catalog No. AA40238A1


Only null left
Explore available promotions

Description

Description

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Strontium molybdenum oxide
Ba < 0.3%
13470-04-7
1 kg
SrMoO4
Strontium molybdate
247.56
(decomposition)
Colorless
Powder
99.9% (excluding Ba)
MFCD00049550
Insoluble in water. Soluble in acid
4.54

Chemical Identifiers

13470-04-7
SrMoO4
MFCD00049550
Strontium molybdate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-730-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only