missing translation for 'onlineSavingsMsg'
Learn More

Strontium molybdenum oxide, 99%

CAS: 13470-04-7 | SrMoO4

$45.18 - $1168.63

Chemical Identifiers

CAS 13470-04-7
Molecular Formula SrMoO4
MDL Number MFCD00049550
Synonym Strontium molybdate
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4023809
View Documents
Thermo Scientific Chemicals
04023809
10 g
Each for $45.18
Only null left
 
AA4023818
View Documents
Thermo Scientific Chemicals
04023818
50 g
Each for $195.95
Only null left
 
AA4023830
View Documents
Thermo Scientific Chemicals
04023830
250 g
Each for $400.73
Only null left
 
AA40238A1
View Documents
Thermo Scientific Chemicals
040238A1
1 kg
Each for $1,168.63
Only null left
 
Description

Description

It is used as an optical material for a small range of special applications, for example, as an optical coating on lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13470-04-7
SrMoO4
MFCD00049550
Strontium molybdate

Specifications

(decomposition)
13470-04-7
10 g
SrMoO4
Strontium molybdate
247.56
Strontium molybdenum oxide, Ba < 0.3%
Colorless
Powder
99.9% (excluding Ba)
MFCD00049550
Insoluble in water. Soluble in acid
4.54
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 236-730-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only