Learn More
Strontium chloride hexahydrate, tech.
CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol
Supplier: Thermo Scientific Chemicals 01249436
Description
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the field of medicine, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Strontium chloride hexahydrate | |
| Crystalline | |
| Cl2H12O6Sr | |
| Soluble in water,alcohol. Slightly soluble in ethanol and acetone. Insoluble in ammonia. | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] | |
| 266.61 | |
| Technical | |
| Hygroscopic |
| 10025-70-4 | |
| 500 g | |
| MFCD00149865 | |
| AMGRXJSJSONEEG-UHFFFAOYSA-L | |
| strontium(2+) hexahydrate dichloride | |
| 266.62 (158.53 Anhydrous) | |
| 1.65 | |
| 1.93 g/mL |
Chemical Identifiers
| 10025-70-4 | |
| 266.61 | |
| AMGRXJSJSONEEG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
| Cl2H12O6Sr | |
| MFCD00149865 | |
| strontium(2+) hexahydrate dichloride |
Safety and Handling
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
EINECSNumber : 233-971-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only