missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride hexahydrate, tech.

Catalog No. p-4407961
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1249436 500 g
AA12494A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1249436 Supplier Thermo Scientific Chemicals Supplier No. 01249436
Only null left

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol

Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the field of medicine, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-70-4
Molecular Formula Cl2H12O6Sr
Molecular Weight (g/mol) 266.61
MDL Number MFCD00149865
InChI Key AMGRXJSJSONEEG-UHFFFAOYSA-L
IUPAC Name strontium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Sr++]

Specifications

Physical Form Crystalline
Quantity 500 g
Solubility Information Soluble in water,alcohol. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
Formula Weight 266.62 (158.53 Anhydrous)
Grade Technical
Refractive Index 1.65
Sensitivity Hygroscopic
Density 1.93 g/mL
Chemical Name or Material Strontium chloride hexahydrate
Hazard Category H318-H335
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 233-971-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only