missing translation for 'onlineSavingsMsg'
Learn More

Strontium Chloride, Hexahydrate, Crystal, ACS, 99.0-103.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S167512KG

 View more versions of this product

Catalog No. 18614109


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10025-70-4
5.0 to 7.0
99 to 103%
Poly Pail
1
12 kg
ACS

Chemical Identifiers

10025-70-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.