missing translation for 'onlineSavingsMsg'
Learn More

Strontium Chloride, Hexahydrate, Crystal, ACS, 99.0-103.0%, Spectrum™ Chemical

Catalog No. p-7166406
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614105 25 g Amber Glass Bottle
18614106 125 g Amber Glass Bottle
18614107 500 g Amber Glass Bottle
18614108 2.5 kg Amber Glass Bottle
18614109 12 kg Poly Pail
18614110 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614105 Supplier Spectrum Chemical Mfg Cor Supplier No. S167525GM
Only null left

Chemical Identifiers

CAS 10025-70-4

Specifications

pH 5.0 to 7.0
CAS Max % 1
Quantity 25 g
Percent Purity 99 to 103%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.