missing translation for 'onlineSavingsMsg'
Learn More

Strontium chloride, anhydrous, 95%

CAS: 10476-85-4

$134.68 - $400.73

Chemical Identifiers

CAS 10476-85-4
MDL Number MFCD00011249
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1220236
View Documents
Thermo Scientific Chemicals
01220236
500 g
Each for $134.68
Only null left
 
AA12202A3
View Documents
Thermo Scientific Chemicals
012202A3
2 kg
Each for $400.73
Only null left
 
Description

Description

It is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10476-85-4
MFCD00011249

Specifications

1250°C
White
Powder
MFCD00011249
Freely soluble in water. Very slightly soluble in ethanol and acetone.
95%
1.65
3.052 g/mL
873°C
10476-85-4
500 g
14,8840
158.53
Odorless
Hygroscopic
Strontium chloride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c

H318-H335

EINECSNumber : 233-971-6

RTECSNumber : WK8400000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only