missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium carbonate, 99%, Ba 1%
CAS: 1633-05-2 | CO3Sr | 147.63 g/mol
$132.92 - $268.33
Chemical Identifiers
| CAS | 1633-05-2 |
|---|---|
| Molecular Formula | CO3Sr |
| Molecular Weight (g/mol) | 147.63 |
| MDL Number | MFCD00011250 |
| InChI Key | LEDMRZGFZIAGGB-UHFFFAOYSA-L |
| Synonym | strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate |
| PubChem CID | 15407 |
| IUPAC Name | strontium;carbonate |
| SMILES | [Sr++].[O-]C([O-])=O |
Description
Used as a colorant in fireworks. It is also used in ceramics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1633-05-2 | |
| 147.63 | |
| LEDMRZGFZIAGGB-UHFFFAOYSA-L | |
| 15407 | |
| [Sr++].[O-]C([O-])=O |
| CO3Sr | |
| MFCD00011250 | |
| strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate | |
| strontium;carbonate |
Specifications
| 1100°C (decomposition) | |
| Powder | |
| 99% | |
| MFCD00011250 | |
| Soluble in ammonium chloride. Slightly soluble in ammonia and water. | |
| [Sr++].[O-]C([O-])=O | |
| 147.63 | |
| 147.63 | |
| Strontium carbonate, 99%, Ba 1% |
| 1633-05-2 | |
| 250 g | |
| CO3Sr | |
| strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate | |
| LEDMRZGFZIAGGB-UHFFFAOYSA-L | |
| strontium;carbonate | |
| 15407 | |
| 3.7 |
Safety and Handling
EINECSNumber : 216-643-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only