missing translation for 'onlineSavingsMsg'
Learn More

Strontium carbonate, 97.5%

CAS: 1633-05-2 | CO3Sr | 147.63 g/mol

$74.51 - $379.53

Chemical Identifiers

CAS 1633-05-2
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
Synonym strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
PubChem CID 15407
IUPAC Name strontium;carbonate
SMILES [Sr++].[O-]C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3339230
View Documents
Thermo Scientific Chemicals
03339230
250 g
Each for $74.51
Only null left
 
AA33392A1
View Documents
Thermo Scientific Chemicals
033392A1
1 kg
Each for $119.88
Only null left
 
AA33392A7
View Documents
Thermo Scientific Chemicals
033392A7
5 kg
Each for $379.53
Only null left
 
Description

Description

As a catalyst, in radiation-resistant glass, in ceramic ferrites, and in pyrotechnicsUsed as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1633-05-2
147.63
LEDMRZGFZIAGGB-UHFFFAOYSA-L
15407
[Sr++].[O-]C([O-])=O
CO3Sr
MFCD00011250
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
strontium;carbonate

Specifications

1100°C (decomposition)
Powder
97.5%
MFCD00011250
Practically insoluble in water. Soluble in acids and carbonated waterSoluble in ammonium chloride. Slightly soluble in ammonia and water.
[Sr++].[O-]C([O-])=O
147.63
147.63
Strontium carbonate
1633-05-2
250 g
CO3Sr
strontium carbonate, strontianite, carbonic acid, strontium salt 1:1, unii-41ypu4mmca, strontium carbonate srco3, ccris 3203, carbonic acid strontium salt 1:1, 41ypu4mmca, strontium, reference standard solution, strontiumcarbonate
LEDMRZGFZIAGGB-UHFFFAOYSA-L
strontium;carbonate
15407
3.7
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-643-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only