missing translation for 'onlineSavingsMsg'
Learn More

Strontium Bromide, Hexahydrate, Purified, 95%, Spectrum™ Chemical

$352.68 - $811.41

Chemical Identifiers

CAS 7789-53-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614094
View Documents
Spectrum Chemical Mfg Cor
S1665125GM
125 g Poly Bottle
Each for $352.68
Only null left
 
18614095
View Documents
Spectrum Chemical Mfg Cor
S1665500GM
500 g Poly Bottle
Each for $811.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7789-53-9

Specifications

7789-53-9
125 g
Purified
1
95%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.