missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptozotocin, ≥97%, MP Biomedicals™
$1,323.18 - $5,822.04
Chemical Identifiers
| CAS | 18883-66-4 |
|---|---|
| Molecular Formula | C8H15N3O7 |
| Molecular Weight (g/mol) | 265.222 |
| InChI Key | ZSJLQEPLLKMAKR-UHEPRFFZSA-N |
| Synonym | STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose |
| PubChem CID | 45357367 |
| IUPAC Name | 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
| SMILES | CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN10055701
|
N/A | 1 g |
Each for $1,323.18
|
|
|||||
|
ICN10055705
|
N/A | 5 g |
Each for $5,822.04
|
|
|||||
Chemical Identifiers
| 18883-66-4 | |
| 265.222 | |
| STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea |
| C8H15N3O7 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 45357367 | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O |
Specifications
| Streptozotocin | |
| 18883-66-4 | |
| ≥97% | |
| STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose | |
| CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O | |
| 265.222 | |
| 265.2 |
| Solid | |
| White | |
| C8H15N3O7 | |
| ZSJLQEPLLKMAKR-UHEPRFFZSA-N | |
| 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea | |
| 45357367 | |
| Streptozotocin |