missing translation for 'onlineSavingsMsg'
Learn More

Streptozotocin, ≥97%, MP Biomedicals™

Catalog No. ICN10055701
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10055701 1 g
ICN10055705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10055701 Supplier MP Biomedicals Inc Supplier No. 0210055701

Chemical Identifiers

CAS 18883-66-4
Molecular Formula C8H15N3O7
Molecular Weight (g/mol) 265.222
InChI Key ZSJLQEPLLKMAKR-UHEPRFFZSA-N
Synonym STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose
PubChem CID 45357367
IUPAC Name 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O

Specifications

Type Streptozotocin
Physical Form Solid
Color White
Quantity 1 g
Assay Percent Range ≥97%
Formula Weight 265.2
Chemical Name or Material Streptozotocin