missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin R PE

Catalog No. SNN1007
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SNN1007 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SNN1007 Supplier Molecular Probes™ Supplier No. SNN1007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin R PE

Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.

Format/Formulation: RPE Conjugate

Specifications

Concentration 0.25 mg/mL
Conjugate R-phycoerythrin
Quantity 500 μg
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Type Streptavidin Conjugate (fluorescent)
How is streptavidin used?

Streptavidin has a high affinity for biotin, with Kd = 1 x 10E-15 moles/L (Green, N.M. [1975] Adv. Protein Chem. 291:85). Streptavidin covalently conjugated to a reporter such as a fluorochrome (R-PE or FITC) or an enzyme (HRP or alkaline phosphatase) has utility in detecting, localizing, and quantitating biotin. Bound streptavidin conjugates can be detected either by fluorescence microscopy or by adding color producing enzyme substrate. This technique is especially useful with biotinylated antibodies. The biotinylation of antibodies is, relatively speaking, an easy procedure and the resulting product will continue to bind to its target antigen with a high degree of specificity and avidity. However, biotin itself cannot be readily detected. Biotin becomes highly apparent once streptavidin conjugated to a reporter is added.

Each streptavidin molecule can bind up to 4 biotin molecules. This feature is potentially useful for cross-linking biotinylated molecules.

For Laboratory Use