missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Streptavidin Protein, HRP

Catalog No. p-7180362
Change view
Click to view available options
Quantity:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21126 1 mg
PI21124 2 mg
PI21127 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI21126 Supplier Thermo Scientific™ Supplier No. 21126
Only null left
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Formulation: 1mg purified protein in PBS and no preservative; pH 6.8
This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures.

Specifications

Conjugate HRP
Quantity 1 mg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Shipping Condition Ambient Temperature
Form Solid
Product Line Pierce
Product Type Streptavidin Conjugate
Target Molecule Biotin
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.