missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin Protein, DyLight™ 800

Catalog No. PI21851
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21851 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21851 Supplier Invitrogen™ Supplier No. 21851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein

21851 contains 1 mL of protein in pale green colored liquid at a concentration of 1 mg/mL.

Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.2
Name Streptavidin
Conjugate DyLight 800
pH Range 7.2
Cross Reactivity Tag
Purification Method Purified
Quantity 1 mL
Storage Requirements 4°C
Regulatory Status RUO
pH 7.2
Common Name Streptavidin
Product Type Protein
Form Liquid
Purity or Quality Grade purified
Host Species N/A
Show More Show Less