missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin Protein, DyLight™ 550
Description
Streptavidin for ICC/IF
Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.
Specifications
Specifications
| Concentration | 1mg/mL |
| Conjugate | DyLight 550 |
| Cross Reactivity | Tag |
| Formulation | Buffered saline solution with 1% BSA and no preservative |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Content And Storage | Stable for 1 year at 4°C |
| Form | Liquid |
| For Use With (Application) | Immunofluorescence |
| Purity or Quality Grade | purified |
| Show More |