missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin Protein, AP

Catalog No. PI21323
Change view
Click to view available options
Quantity:
3 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21323 3 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21323 Supplier Invitrogen™ Supplier No. LSG21323

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein

Streptavidin for Western Blot, ELISA

Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.

Specifications

Concentration 2 to 4 mg/mL
For Use With (Application) ELISA, Western Blot
Formulation TBS with 50% glycerol, 0.1mM MgCl2, 0.1mM ZnCl2 and no preservative; pH 8
Name Streptavidin
Conjugate Alkaline Phosphatase (AP)
pH Range 8
Cross Reactivity Tag
Purification Method Purified
Quantity 3 mg
Storage Requirements -20°C
Regulatory Status RUO
pH 8
Common Name Streptavidin
Product Type Protein
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Form Liquid
Purity or Quality Grade purified
Host Species N/A
Show More Show Less