missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate

Catalog No. 5016042
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5016042 25 μg
5016043 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5016042 Supplier Invitrogen™ Supplier No. 46431780

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Blue Laser

Emit

710 nm

Excite

488 nm

Reported Application

Flow Cytometric Analysis

Specifications

Concentration 0.2 mg/mL
Conjugate PerCP-eFluor 710
Quantity 25 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PerCP-eFluor 710

For Research Use Only