missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate

Catalog No. p-7091861
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
25 μg
Catalog No. Quantity
5015835 25 μg
5015836 100 μg
2 options
Contains Sodium azide

Catalog No. 5015835

Supplier: Invitrogen™ 45431780

Only null left
Contains Sodium azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Specifications

Concentration 0.2 mg/mL
Conjugate PerCP-Cy5.5
Quantity 25 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PerCP-Cy5.5

For Research Use Only