missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (PE-TR)

Catalog No. 501121552
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121552 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121552 Supplier Invitrogen™ Supplier No. SA1017

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, (PE-TR)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Concentration 1 mg/mL
Conjugate PE-Texas Red
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 0.5 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, PE-TR conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/615
Flow Cytometer Laser Lines 488 or 561, 488
Shipping Condition Wet Ice
Form Solution
Product Line Texas Red
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye PE-Texas Red
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.