missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (PE-Cy7)

Catalog No. 501137616
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137616 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137616 Supplier Invitrogen™ Supplier No. SA1012

Streptavidin, (PE-Cy7)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Concentration 1 mg/mL
Conjugate PE-Cy7
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 0.5 mL
Content And Storage Contains 1 vial of streptavidin,PE-Cy7 conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/767
Flow Cytometer Laser Lines 488, 488 or 561, 561
Shipping Condition Wet Ice
Form Solution
Product Line Cy
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye PE-Cy7
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.