missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptavidin - HRP
Description
Reactivity: HRP-SA reacts with biotin.
Applications: Immunohistochemistry, Western blotting, and ELISA
Specifications
Specifications
| Concentration | 1.25 mg/mL |
| Conjugate | HRP |
| Quantity | 2.5 mg |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Wet Ice |
| Form | Liquid |
| Product Type | Streptavidin Conjugate (fluorescent) |
| Label or Dye | HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.