missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 710 Conjugate

Catalog No. 5016424
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5016424 100 μg
5016423 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Azide
Catalog No. 5016424 Supplier Invitrogen™ Supplier No. 49431782

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Red Laser

Emit

710 nm

Excite

633 - 647 nm

Reported Application

Flow Cytometric Analysis

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 710
Quantity 100 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye eFluor 710

For Research Use Only