missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 450 Conjugate

Catalog No. 501128802
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501128802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Azide
Catalog No. 501128802 Supplier Invitrogen™ Supplier No. 48431782

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

Laser

Violet Laser

Emit

445 nm

Excite

405 nm

Reported Application

Flow Cytometric Analysis

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 450
Quantity 100 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye eFluor 450

For Research Use Only. Not for use in diagnostic procedures.