missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin - Cy5

Catalog No. 434316
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
434316 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 434316 Supplier Invitrogen™ Supplier No. 434316

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin - Cy5

Reactivity: CY5-SA reacts with biotin.

Specifications

Concentration 1 mg/mL
Conjugate Cy5
Quantity 1 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Line Cy
Product Type Streptavidin Conjugate (fluorescent)
Is there a crosslinker used in Streptavidin-FITC (Cat. No. 434311) and Streptavidin-Cy5 (Cat. No. 434316) conjugates?

There is no crosslinker as the dyes themselves are made to react directly with the streptavidin protein.

How many molecules of protein and dye are in 1 mg of Streptavidin-Cy5 (Cat. No. 434316)?

There are about 3.993 x 10^16 molecules of dye and 1.141 x 10^16 of streptavidin in 1 mg of Streptavidin-Cy5.

What is the dye-protein ratio for Streptavidin-Cy5 (Cat. No. 434316)?

The target dye to protein ratio for Streptavidin-Cy5 (Cat. No. 434316) is 4:1. The average number of dye molecules on each streptavidin molecule is between 3 and 4.

For Research Use Only. Not for use in diagnostic procedures.