missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (Cy5)

Catalog No. 501121551
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121551 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121551 Supplier Invitrogen™ Supplier No. LSSA1011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, (Cy5)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Conjugate Cy5
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 0.5 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, Cy5 conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 600-650/670
Flow Cytometer Laser Lines 633/635
Shipping Condition Wet Ice
Form Solution
Product Line Cy
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Cy5
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.