missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin-CY3 (ZyMax Grade)

Catalog No. 438315
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
438315 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 438315 Supplier Invitrogen™ Supplier No. 438315

Streptavidin-CY3 (ZyMax Grade)

Applications: Immunohistochemistry, Western blotting, ELISA, ISH

ZyMAX™ reagents offer enhanced assay sensitivity, higher titer, and greater flexibility in assay design than standard grade reagents.

Specifications

Concentration 1 mg/mL
Conjugate Cy3
Quantity 1 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid
Product Line Invitrogen
Product Type Streptavidin Conjugate (fluorescent)

For Research Use Only. Not for use in diagnostic procedures.