missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptavidin, (Cy3)

Catalog No. 501121550
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121550 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121550 Supplier Invitrogen™ Supplier No. SA1010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, (Cy3)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide

Specifications

Conjugate Cy3
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Quantity 0.5 mL
Packaging Tube
Content And Storage Contains 1 vial of streptavidin, Cy3 conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/565
Flow Cytometer Laser Lines 488
Shipping Condition Wet Ice
Form Solution
Product Line Cy
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Cy3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.